Learn More
ATCC Proteus penneri; Strain CDC 1808-73 N
Supplier:  ATCC 33519
Encompass
Proteus penneri strain CDC 1808-73 was isolated in Anchorage, Alaska, from a urine sample. This bacterial type strain grows aerobically in nutrient agar/broth.
Catalog No. 50-237-9904