Learn More
Creative Biomart Recombinant Treponema pallidum p17 Antigen, His-tagged
Catalog No. 502084816
Encompass_Preferred
Recombinant Treponema pallidum p17 derived from full length TP17 gene containing 139 aa produced in E. coli and fused to a 6 His tag at C-terminus. The total Mw is 22 kDa and the pI is 8.7, was pur
Catalog No. 50-208-4816 Supplier Creative Biomart Supplier No. TPP438
Recombinant Treponema pallidum p17 derived from full length TP17 gene containing 139 aa produced in E. coli and fused to a 6 His tag at C-terminus. The total Mw is 22 kDa and the pI is 8.7, was pur