Learn More
Creative Biomart Recombinant Treponema p47 Antigen, His-tagged
Supplier:  Creative Biomart TPP439
Encompass_Preferred
Recombinant Treponema p47 produced in E. coli is a non-glycosylated polypeptide chain having a molecular mass of 47.7 kDa and fused to a His tag at N-terminus.
Catalog No. 50-208-4817