Learn More
Diagnostic Biosystems TREPONEMA PALLIDUM Treponema Pallidum
Catalog No. 502407477
Encompass_Preferred
Treponema pallidum is the causative agent of syphilis. It is a gram negative spirochete, a helical to sinusoidal bacterium with outer and cytoplasmic membranes, a thin peptidoglycan layer, and periplasmic flagella.
Catalog No. 50-240-7477 Supplier Diagnostic Biosystems Supplier No. PDM577
Treponema pallidum is the causative agent of syphilis. It is a gram negative spirochete, a helical to sinusoidal bacterium with outer and cytoplasmic membranes, a thin peptidoglycan layer, and periplasmic flagella.