Learn More
Bioworld Treponema pallidum TmpA (Partial) (Recombinant), 100 μg
Catalog No. 50253832
Encompass_Preferred
Recombinant Treponema Pallidum Membrane Protein A (23-41aa, 288-325aa) Source: E.coli Description : The protein contains the Trp. Pallidum Membrane Protein A immunodominant regions (23-41aa, 288-325aa). Purification method: Purified by proprietary chromatographic technique. Purity: Protein is >90% pure as determined by 10% PAGE (coomassie staining).. Protein conc: 1mg/ml, 10mM Tris-HCl pH8.0, 1mM EDTA, 20mMDTT and 8M urea. Storage: Protein is shipped at ambient temperature. Upon arrival, Store at -20C. Stability: Five years frozen. One month in solution at room temperature. Specificity: Immunoreactive with sera of Trp. Pallidum infected individuals. Application: Antigen in ELISA and Western blots, excellent antigen for detection of Trp. Pallidum with minimal specificity problems. Usage: BioWORLD\'s products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-832 Supplier Bioworld Supplier No. 220607341
Recombinant Treponema Pallidum Membrane Protein A (23-41aa, 288-325aa) Source: E.coli Description : The protein contains the Trp. Pallidum Membrane Protein A immunodominant regions (23-41aa, 288-325aa). Purification method: Purified by proprietary chromatographic technique. Purity: Protein is >90% pure as determined by 10% PAGE (coomassie staining).. Protein conc: 1mg/ml, 10mM Tris-HCl pH8.0, 1mM EDTA, 20mMDTT and 8M urea. Storage: Protein is shipped at ambient temperature. Upon arrival, Store at -20C. Stability: Five years frozen. One month in solution at room temperature. Specificity: Immunoreactive with sera of Trp. Pallidum infected individuals. Application: Antigen in ELISA and Western blots, excellent antigen for detection of Trp. Pallidum with minimal specificity problems. Usage: BioWORLD\'s products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.