Learn More
Creative Biomart Recombinant Treponema TmpA Antigen, His-tagged
Supplier:  Creative Biomart TMP440
Encompass_Preferred
Recombinant Treponema TmpA produced in E. coli is a non-glycosylated polypeptide chain having a molecular mass of 45 kDa and fused to a His tag at N-terminus.
Catalog No. 50-208-4818