Learn More
Medchemexpress LLC HY-100441 2mg Medchemexpress, Treprostinil CAS:81846-19-7 Purity:>98%
SDP
Catalog No. 501931836
Encompass_Preferred
Medchemexpress, HY-100441 2mg Treprostinil CAS:81846-19-7 Treprostinil (LRX-15) is a potent DP1 and EP2 agonist with EC 50 values of 0.6±0.1 and 6.2±1.2 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1836 Supplier Medchemexpress LLC Supplier No. HY1004412MG
Medchemexpress, HY-100441 2mg Treprostinil CAS:81846-19-7 Treprostinil (LRX-15) is a potent DP1 and EP2 agonist with EC 50 values of 0.6±0.1 and 6.2±1.2 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.