Learn More
Medchemexpress LLC Treprostinil | 81846-19-7 | 390.51 g/mol | C23H34O5 | 50 MG
SDP
Catalog No. 5000214908
Encompass_Preferred
Treprostinil is a synthetic prostacyclin analogue supplied as a research reagent. It is a potent agonist of prostanoid receptors DP1 and EP2, with EC50 values of 0.6 ± 0.1 nM (DP1) and 6.2 ± 1.2 nM (EP2). The compound is provided as a powder for laboratory use; powder storage at -20°C, with in-solution stability of -80°C for 6 months or -20°C for 1 month.
  • Potent DP1 and EP2 agonist with EC50s of 0.6 ± 0.1 nM (DP1) and 6.2 ± 1.2 nM (EP2).
  • Supplied as a powder for laboratory research and storage.
  • Molecular formula C23H34O5 and molecular weight 390.51 g/mol.
  • Store powder at -20°C; solutions stable at -80°C for 6 months or -20°C for 1 month.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-14908 Supplier Medchemexpress LLC Supplier No. HY10044150MG
Treprostinil is a synthetic prostacyclin analogue supplied as a research reagent. It is a potent agonist of prostanoid receptors DP1 and EP2, with EC50 values of 0.6 ± 0.1 nM (DP1) and 6.2 ± 1.2 nM (EP2). The compound is provided as a powder for laboratory use; powder storage at -20°C, with in-solution stability of -80°C for 6 months or -20°C for 1 month.
  • Potent DP1 and EP2 agonist with EC50s of 0.6 ± 0.1 nM (DP1) and 6.2 ± 1.2 nM (EP2).
  • Supplied as a powder for laboratory research and storage.
  • Molecular formula C23H34O5 and molecular weight 390.51 g/mol.
  • Store powder at -20°C; solutions stable at -80°C for 6 months or -20°C for 1 month.
  • For research use only; not for human or veterinary use.