Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Treprostinil-d9 | 2747918-14-3 | 99.8% | 399.57 g/mol | C23H25D9O5 | 10 MG
SDP

Catalog No. 5000213747
Encompass_Preferred

Treprostinil-d9 is the deuterium-labeled (D9) isotopologue of treprostinil supplied as an analytical standard and internal tracer for quantitative assays. It is intended for research use only and supports method development and accurate bioanalytical quantitation.

  • Deuterium-labeled internal standard for quantitative assays.
  • Suitable for NMR, GC-MS, and LC-MS analysis.
  • High purity (99.8%) for analytical accuracy.
  • Molecular weight 399.57 g/mol; formula C23H25D9O5.
  • Available in small-mass packages, including 10 MG quantities.
  • For research use only; not for human or therapeutic use.

Catalog No. 50-002-13747 Supplier Medchemexpress LLC Supplier No. HY100441S210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Treprostinil-d9 is the deuterium-labeled (D9) isotopologue of treprostinil supplied as an analytical standard and internal tracer for quantitative assays. It is intended for research use only and supports method development and accurate bioanalytical quantitation.

  • Deuterium-labeled internal standard for quantitative assays.
  • Suitable for NMR, GC-MS, and LC-MS analysis.
  • High purity (99.8%) for analytical accuracy.
  • Molecular weight 399.57 g/mol; formula C23H25D9O5.
  • Available in small-mass packages, including 10 MG quantities.
  • For research use only; not for human or therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.