Learn More
Medchemexpress LLC Treprostinil-d9 | 2747918-14-3 | MFCD23144256 | 99.8% | 399.57 g/mol | C23H25D9O5 | 5 MG
SDP
Catalog No. 5000215076
Encompass_Preferred
Treprostinil-d9 is a deuterium-labeled analogue of treprostinil used as a stable-isotope internal standard for analytical and pharmacokinetic studies. It is intended for use in mass spectrometry, method development, and quantitative bioanalysis where accurate normalization and peak identification are required.
  • Deuterium-labeled internal standard for accurate quantitation in MS assays.
  • High purity (99.79%) for reliable analytical performance.
  • Molecular weight approximately 399.57 g/mol for precise mass calibration.
  • Available in small, research-friendly pack sizes (mg scale).
  • Suitable for pharmacokinetic and bioanalytical method development.
Catalog No. 50-002-15076 Supplier Medchemexpress LLC Supplier No. HY100441S25MG
Treprostinil-d9 is a deuterium-labeled analogue of treprostinil used as a stable-isotope internal standard for analytical and pharmacokinetic studies. It is intended for use in mass spectrometry, method development, and quantitative bioanalysis where accurate normalization and peak identification are required.
  • Deuterium-labeled internal standard for accurate quantitation in MS assays.
  • High purity (99.79%) for reliable analytical performance.
  • Molecular weight approximately 399.57 g/mol for precise mass calibration.
  • Available in small, research-friendly pack sizes (mg scale).
  • Suitable for pharmacokinetic and bioanalytical method development.