Learn More
Medchemexpress LLC Treprostinil-d9 (UT-15-d9) | 2747918-14-3 | MFCD23144256 | 99.8% | 399.57 | C23H25D9O5 | 1 MG
SDP
Catalog No. 5000215351
Encompass_Preferred
Treprostinil-d9 (UT-15-d9) is a deuterium-labeled analogue of treprostinil provided for research and analytical applications. It functions as a tracer and internal standard for quantitative assays, and mirrors the parent compound's activity at prostanoid receptors.
  • Used as an internal standard for quantitative NMR, GC-MS, and LC-MS
  • Deuterium-labeled tracer suitable for pharmacokinetic and metabolic studies
  • Potent DP1 and EP2 receptor agonist activity (parent compound reported EC50 values)
  • High purity for analytical applications
  • Solid powder with defined cold storage stability
  • Intended for research use only
Catalog No. 50-002-15351 Supplier Medchemexpress LLC Supplier No. HY100441S21MG
Treprostinil-d9 (UT-15-d9) is a deuterium-labeled analogue of treprostinil provided for research and analytical applications. It functions as a tracer and internal standard for quantitative assays, and mirrors the parent compound's activity at prostanoid receptors.
  • Used as an internal standard for quantitative NMR, GC-MS, and LC-MS
  • Deuterium-labeled tracer suitable for pharmacokinetic and metabolic studies
  • Potent DP1 and EP2 receptor agonist activity (parent compound reported EC50 values)
  • High purity for analytical applications
  • Solid powder with defined cold storage stability
  • Intended for research use only