Learn More
TARGETMOL CHEMICALS INC TREPROSTINIL DIETHANOLAM 5MG
SDP
Catalog No. 502579426
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Treprostinil diethanolamine (UT-15C) is a potent agonist of EP2 DP1 and IP with values of 3.6 4.4 32.1 212 826 2505 and 4680 nM for EP2 DP1 IP EP1 EP4 EP3 and FPKi respectively. Treprostinil diethanolamine is capable of contributing to the upregulation of cAMP which in turn maintains homeostasis within the vascular system and causes vasodilation in human pulmonary arteries. purity: 98%
Catalog No. 50-257-9426 Supplier TARGETMOL CHEMICALS INC Supplier No. T633495MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Treprostinil diethanolamine (UT-15C) is a potent agonist of EP2 DP1 and IP with values of 3.6 4.4 32.1 212 826 2505 and 4680 nM for EP2 DP1 IP EP1 EP4 EP3 and FPKi respectively. Treprostinil diethanolamine is capable of contributing to the upregulation of cAMP which in turn maintains homeostasis within the vascular system and causes vasodilation in human pulmonary arteries. purity: 98%