Learn More
Medchemexpress LLC Treprostinil sodium | 289480-64-4 | 99.55% | 412.49 | 1 ML
SDP
Catalog No. 5000391663
Encompass_Preferred
Treprostinil (UT-15) sodium is a potent DP1 and EP2 agonist. It exhibits high affinity for the DP1, EP2, and IP receptors, with lower affinity for EP1, EP4, EP3, FP, and TP receptors. Activation of IP, DP1, and EP2 receptors can lead to vasodilation of human pulmonary arteries.
  • Potent DP1 and EP2 agonist.
  • High affinity for DP1, EP2, and IP receptors.
  • Can lead to vasodilation of human pulmonary arteries.
  • Inhibits the viability of cultured endothelial colony-forming cells.
Catalog No. 50-003-91663 Supplier Medchemexpress LLC Supplier No. HY1650410MM/1ML
Treprostinil (UT-15) sodium is a potent DP1 and EP2 agonist. It exhibits high affinity for the DP1, EP2, and IP receptors, with lower affinity for EP1, EP4, EP3, FP, and TP receptors. Activation of IP, DP1, and EP2 receptors can lead to vasodilation of human pulmonary arteries.
  • Potent DP1 and EP2 agonist.
  • High affinity for DP1, EP2, and IP receptors.
  • Can lead to vasodilation of human pulmonary arteries.
  • Inhibits the viability of cultured endothelial colony-forming cells.