Learn More
Medchemexpress LLC Treprostinil (UT-15) | 81846-19-7 | 100.0% | 390.51 | C23H34O5 | 100 MG
SDP
Catalog No. 5000214712
Encompass_Preferred
Treprostinil (UT-15) is a prostacyclin analog that functions as a potent DP1 and EP2 agonist, provided for research and analytical applications. The substance is accompanied by a manufacturer datasheet and analytical documentation for experimental use.
  • Potent DP1 and EP2 agonist with low-nanomolar EC50s (0.6 and 6.2 nM).
  • Suitable for receptor pharmacology and in vitro assays.
  • Available as solid or as a 10 mM solution in DMSO for easy reconstitution.
  • High purity (99.95-99.98%) for analytical use.
  • Documented with datasheet, COA, SDS, and analytical spectra.
Catalog No. 50-002-14712 Supplier Medchemexpress LLC Supplier No. HY100441100MG
Treprostinil (UT-15) is a prostacyclin analog that functions as a potent DP1 and EP2 agonist, provided for research and analytical applications. The substance is accompanied by a manufacturer datasheet and analytical documentation for experimental use.
  • Potent DP1 and EP2 agonist with low-nanomolar EC50s (0.6 and 6.2 nM).
  • Suitable for receptor pharmacology and in vitro assays.
  • Available as solid or as a 10 mM solution in DMSO for easy reconstitution.
  • High purity (99.95-99.98%) for analytical use.
  • Documented with datasheet, COA, SDS, and analytical spectra.