Learn More
Enzo Life Sciences Trequinsin (10mg). CAS: 78416-81-6
SDP
Supplier:  Enzo Life Sciences BMLPD1800010
Encompass_Preferred
Ultrapotent, highly selective inhibitor of cGMP-inhibited phosphodiesterase (PDE lll, IC50=0.3nM) in vitro. Potentiates adenosine-stimulated cAMP accumulation. Alternative name: 9,10-Dimethoxy-2-mesitylimino-3-methyl-2,3,6,7-tetrahydro-4H-pyrimido-[6,1a]-isoquinolin-4-one . HCl, HL 725. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (20mg/ml); slightly soluble in water. Long Term Storage: Ambient.
Catalog No. 50-201-1391