Learn More
Medchemexpress LLC Trequinsin hydrochloride | 78416-81-6 | 99.9% | 441.95 | 25 MG
SDP
Catalog No. 5000384480
Encompass_Preferred
Trequinsin hydrochloride (HL 725) is a highly potent inhibitor of platelet CAMP phosphodiesterase (PDE), with an IC50 of 0.25 nM. It is also a very potent inhibitor of the aggregation of human platelets induced in vitro by ADP, collagen, thrombin, and epinephrine. In addition to its cardiovascular and antihypertensive qualities, it exhibits very potent antiplatelet activities. It is also an efficacious agonist of [Ca2+]i.
  • Extremely potent inhibitor of platelet CAMP phosphodiesterase (PDE)
  • Inhibits aggregation of human platelets induced by ADP, collagen, thrombin, and epinephrine
  • Possesses cardiovascular and antihypertensive qualities
  • Exhibits potent antiplatelet activities
  • Efficacious agonist of [Ca2+]i
Catalog No. 50-003-84480 Supplier Medchemexpress LLC Supplier No. 25MGHY18740A
Trequinsin hydrochloride (HL 725) is a highly potent inhibitor of platelet CAMP phosphodiesterase (PDE), with an IC50 of 0.25 nM. It is also a very potent inhibitor of the aggregation of human platelets induced in vitro by ADP, collagen, thrombin, and epinephrine. In addition to its cardiovascular and antihypertensive qualities, it exhibits very potent antiplatelet activities. It is also an efficacious agonist of [Ca2+]i.
  • Extremely potent inhibitor of platelet CAMP phosphodiesterase (PDE)
  • Inhibits aggregation of human platelets induced by ADP, collagen, thrombin, and epinephrine
  • Possesses cardiovascular and antihypertensive qualities
  • Exhibits potent antiplatelet activities
  • Efficacious agonist of [Ca2+]i