Learn More
Medchemexpress LLC Trequinsin hydrochloride | 78416-81-6 | 99.9% | 441.95 | 5 MG
SDP
Catalog No. 5000387961
Encompass_Preferred
Trequinsin hydrochloride (HL 725) is an extremely potent inhibitor of platelet cAMP phosphodiesterase (PDE) with an IC50 of 0.25 nM. It is also an extremely potent inhibitor of the aggregation of human platelets induced in vitro by ADP, collagen, thrombin and epinephrine. Trequinsin hydrochloride exerts very potent antiplatelet activities and is an efficacious agonist of [Ca2+]i.
  • Extremely potent inhibitor of platelet cAMP phosphodiesterase (PDE) with an IC50 of 0.25 nM.
  • Efficacious agonist of [Ca2+]i.
  • Inhibits aggregation of human platelets induced by ADP, collagen, thrombin, and epinephrine in vitro.
  • Appearance: solid.
  • Color: light yellow to yellow.
Catalog No. 50-003-87961 Supplier Medchemexpress LLC Supplier No. HY18740A5MG
Add to Cart
Add to Cart
Trequinsin hydrochloride (HL 725) is an extremely potent inhibitor of platelet cAMP phosphodiesterase (PDE) with an IC50 of 0.25 nM. It is also an extremely potent inhibitor of the aggregation of human platelets induced in vitro by ADP, collagen, thrombin and epinephrine. Trequinsin hydrochloride exerts very potent antiplatelet activities and is an efficacious agonist of [Ca2+]i.
  • Extremely potent inhibitor of platelet cAMP phosphodiesterase (PDE) with an IC50 of 0.25 nM.
  • Efficacious agonist of [Ca2+]i.
  • Inhibits aggregation of human platelets induced by ADP, collagen, thrombin, and epinephrine in vitro.
  • Appearance: solid.
  • Color: light yellow to yellow.