Learn More
Medchemexpress LLC Trequinsin hydrochloride | 78416-81-6 | MFCD01076563 | 99.86% | 441.95 | 50 MG
SDP
Catalog No. 5000387960
Encompass_Preferred
Trequinsin hydrochloride (HL 725) is an extremely potent inhibitor of platelet CAMP phosphodiesterase (PDE), with an IC50 of 0.25 nM. It is also an extremely potent inhibitor of the aggregation of human platelets induced in vitro by ADP, collagen, thrombin, and epinephrine. Trequinsin hydrochloride exerts potent antiplatelet activities and is an efficacious agonist of [Ca2+]i.
  • Extremely potent inhibitor of platelet CAMP phosphodiesterase (PDE)
  • Inhibits aggregation of human platelets induced by ADP, collagen, thrombin, and epinephrine
  • Exerts potent antiplatelet activities
  • Efficacious agonist of [Ca2+]i
Catalog No. 50-003-87960 Supplier Medchemexpress LLC Supplier No. HY18740A50MG
Add to Cart
Add to Cart
Trequinsin hydrochloride (HL 725) is an extremely potent inhibitor of platelet CAMP phosphodiesterase (PDE), with an IC50 of 0.25 nM. It is also an extremely potent inhibitor of the aggregation of human platelets induced in vitro by ADP, collagen, thrombin, and epinephrine. Trequinsin hydrochloride exerts potent antiplatelet activities and is an efficacious agonist of [Ca2+]i.
  • Extremely potent inhibitor of platelet CAMP phosphodiesterase (PDE)
  • Inhibits aggregation of human platelets induced by ADP, collagen, thrombin, and epinephrine
  • Exerts potent antiplatelet activities
  • Efficacious agonist of [Ca2+]i