Learn More
Apexbio Technology LLC Tretinoin (Aberela), 100mg Cas# 302-79-4
SDP
Catalog No. 501998598
Encompass_Preferred
Tretinoin (also known as all-trans retinoic acid or ATRA) is an active retinoid metabolite form of Vitamin A. 302-79-4. MFCD00001551 Tretinoin(Aberela) is retinoic acid in pharmaceutical form and used to treat acne vulgaris and keratosis pilaris. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8598 Supplier Apexbio Technology LLC Supplier No. A8539100
Tretinoin (also known as all-trans retinoic acid or ATRA) is an active retinoid metabolite form of Vitamin A. 302-79-4. MFCD00001551 Tretinoin(Aberela) is retinoic acid in pharmaceutical form and used to treat acne vulgaris and keratosis pilaris. There are other sizes available. Please inqury us for quote.