Learn More
Apexbio Technology LLC Tretinoin (Aberela), 5g Cas# 302-79-4
SDP
Supplier:  Apexbio Technology LLC A85395000
Encompass_Preferred
Tretinoin (also known as all-trans retinoic acid or ATRA) is an active retinoid metabolite form of Vitamin A. 302-79-4. MFCD00001551 Tretinoin(Aberela) is retinoic acid in pharmaceutical form and used to treat acne vulgaris and keratosis pilaris. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8601