Learn More
MSC Magid T-REX Impact Gloves, Foam Nitrile, Cut A4, 1 ea.
Catalog No. 5000688664
Encompass
These low-profile impact gloves are engineered for comfort, cut resistance, and performance in light-duty environments. The 13-gauge, seamless machine-knit shell is constructed from Hyperon® fiber, providing breathability and ANSI Level A4 cut protection. Utilizing the M-Force Defense System®, the gloves feature ultra-flexible, low-profile thermoplastic rubber (TPR) pads on the back of the hand and fingers to absorb impacts while maintaining dexterity. A foam nitrile palm coating delivers superior grip in liquid and light oil applications, along with high levels of puncture and abrasion resistance.
  • ANSI Level A4 cut, Level 4 abrasion, and Level 4 puncture resistance
  • ANSI/ISEA 138 Impact Level 1 protection on knuckles and fingers
  • Forty percent lighter than a standard impact glove
  • Touchscreen compatible design
  • Flat, nitrile-reinforced thumb saddle
  • Machine washable
Catalog No. 50-006-88664 Supplier MSC Supplier No. 93758423
These low-profile impact gloves are engineered for comfort, cut resistance, and performance in light-duty environments. The 13-gauge, seamless machine-knit shell is constructed from Hyperon® fiber, providing breathability and ANSI Level A4 cut protection. Utilizing the M-Force Defense System®, the gloves feature ultra-flexible, low-profile thermoplastic rubber (TPR) pads on the back of the hand and fingers to absorb impacts while maintaining dexterity. A foam nitrile palm coating delivers superior grip in liquid and light oil applications, along with high levels of puncture and abrasion resistance.
  • ANSI Level A4 cut, Level 4 abrasion, and Level 4 puncture resistance
  • ANSI/ISEA 138 Impact Level 1 protection on knuckles and fingers
  • Forty percent lighter than a standard impact glove
  • Touchscreen compatible design
  • Flat, nitrile-reinforced thumb saddle
  • Machine washable