Learn More
Aviva Systems Biology Corp TRF3 Antibody - N-terminal region (ARP37625_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37625_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TRF3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse TRF3. Alternate names: Trf3, Gm348, RP23-263E15.2ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-014-53084