Learn More
Sigma Aldrich Fine Chemicals Biosciences Trfluoroacetc acd purs250ML
Catalog No. 502907937
Encompass
Trifluoroacetic acid (TFA) is a fluorinated acetic acid. It is widely used as trifluoromethylating reagent for trifluoromethylation of aromatic compounds.
Catalog No. 50-290-7937 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 91707250ML
Trifluoroacetic acid (TFA) is a fluorinated acetic acid. It is widely used as trifluoromethylating reagent for trifluoromethylation of aromatic compounds.