Learn More
Cayman Chemical TRFRET asy BuFR AddItIve 200mg
SDP
Catalog No. 502896011
Encompass_Preferred
A quinolone antibiotic active against clinical isolates of S. pyogenes P aerUGinosa and P. acnes and clinical isolates of antibiotic-sensitive and -resistant S. aureus S. epidermidis and S. pneumoniae (MIC50s 0.00625-1.56 mg/L) inhibits S. aureus DNA gyrase and topoisomerase IV (IC50s 4.5 and 0.617 mg/L respectively) reduces insulin- or 5a-dihydroxy testosterone-induced triglyceride production and lipid droplet formation in primary hamster sebocytes at 30 UM decreases heat-killed C. acnes-induced increases in IL-6 and IL-8 production in HEKa primary human keratinocytes and IL-1B IL-6 IL-8 and TNF-a production in THP-1 monocytes at 30 UG/ml decreases skin lesion bacterial counts in a mouse model of burn infection caused by S. aureus
Catalog No. 50-289-6011 Supplier Cayman Chemical Supplier No. 600504200 MG
A quinolone antibiotic active against clinical isolates of S. pyogenes P aerUGinosa and P. acnes and clinical isolates of antibiotic-sensitive and -resistant S. aureus S. epidermidis and S. pneumoniae (MIC50s 0.00625-1.56 mg/L) inhibits S. aureus DNA gyrase and topoisomerase IV (IC50s 4.5 and 0.617 mg/L respectively) reduces insulin- or 5a-dihydroxy testosterone-induced triglyceride production and lipid droplet formation in primary hamster sebocytes at 30 UM decreases heat-killed C. acnes-induced increases in IL-6 and IL-8 production in HEKa primary human keratinocytes and IL-1B IL-6 IL-8 and TNF-a production in THP-1 monocytes at 30 UG/ml decreases skin lesion bacterial counts in a mouse model of burn infection caused by S. aureus