Learn More
Bachem TRH
SDP
Supplier:  Bachem 4038214.0250
Encompass_Preferred
TRH , Pyr-His-Pro-NH2, 24305-27-9, C16H22N6O4, Mr 362.39, 250mg. Prolactoliberin, Thyroliberin, Prolactostatin, Protirelin, Thyrotropin-Releasing Hormone. TRH exhibits a variety of biological effects, when injected into humans or animals. Its physiological role appears to be the stimulation of thyrotropin (TSH) and prolactin (PRL) release from the anterior pituitary. Though it was first identified in the hypothalamus as a regulator of the pituitary-thyroid axis, TRH was detected as well by Luo et al. in pancreatic b-cells co-localized with insulin. Application of the hormone reverted hyperglycemia in rats. Moreover, TRH plays an important role in the control of human hair-growth and hair follicle pigmentation.
Catalog No. 50-259-894