Learn More
Aviva Systems Biology Corp TRH Antibody - C-terminal region (AVARP13110_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13110_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TRH. Alternate names: MGC125964, MGC125965ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Zebrafish
Catalog No. 50-014-62644