Learn More
Aviva Systems Biology Corp TRHR Antibody - middle region (AVARP13104_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13104_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TRHR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRHR. Alternate names: TRH-RApplicationWB. Species reactivity: Dog, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62642