Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri(1-naphthyl)phosphine, 97%

Catalog No. AA4230418
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
50 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4230418 50 g
AA4230402 0.5 g
AA4230404 2 g
AA4230409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA4230418 Supplier Thermo Scientific Chemicals Supplier No. 04230418
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3411-48-1 | C30H21P | 412.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3411-48-1
Molecular Formula C30H21P
Molecular Weight (g/mol) 412.47
MDL Number MFCD00046400
InChI Key DMEUUKUNSVFYAA-UHFFFAOYSA-N
Synonym tri 1-naphthyl phosphine, tri-1-naphthylphosphine, tri naphthalen-1-yl phosphine, tris 1-naphthyl phosphine, tris naphthalen-1-yl phosphane, trinaphthyl phosphine, tri-1-naphtylphosphine, tri-1-napthylphosphine, acmc-1cp9e, tri naphthalen-1-yl phosphane
PubChem CID 271443
IUPAC Name trinaphthalen-1-ylphosphane
SMILES C1=CC2=CC=CC(P(C3=CC=CC4=CC=CC=C34)C3=C4C=CC=CC4=CC=C3)=C2C=C1

Specifications

Melting Point 265°C to 268°C
Quantity 50 g
Formula Weight 412.47
Physical Form Powder
Chemical Name or Material Tri (1-naphthyl)phosphine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 627-667-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.