Learn More
Strem, An Ascensus Company TRI-2-FURYLPHOSPHINE 98+%-2G
SDP
Catalog No. 5090110421
Encompass
Tri-2-furylphosphine, 98+%; Size: 2g; [5518-52-5]; (C4H3O)3P; Mol Wt:232.17; Color & Form: white pwdr.
Catalog No. 50-901-10421 Supplier Strem, An Ascensus Company Supplier No. 1563722G
Tri-2-furylphosphine, 98+%; Size: 2g; [5518-52-5]; (C4H3O)3P; Mol Wt:232.17; Color & Form: white pwdr.