Learn More
Strem, An Ascensus Company CAS# 5518-52-5. 5G. Tri-2-furylphosphine, 98+%. MFCD00151857
SDP
Catalog No. 501963594
Encompass
CAS# 5518-52-5. 5G. Tri-2-furylphosphine, 98+%. MFCD00151857. Molecular Formula: (C4H3O)3P. Molecular Weight: 232.17. Color/Form: white pwdr. Stability: air sensitive. Strem# 15-6272. www.strem.com/catalog/v3/15-6372/
Catalog No. 50-196-3594 Supplier Strem, An Ascensus Company Supplier No. 1563725G
CAS# 5518-52-5. 5G. Tri-2-furylphosphine, 98+%. MFCD00151857. Molecular Formula: (C4H3O)3P. Molecular Weight: 232.17. Color/Form: white pwdr. Stability: air sensitive. Strem# 15-6272. www.strem.com/catalog/v3/15-6372/