Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tri(2-methylphenyl)phosphine | 6163-58-2 | 99.99% | C21H21P | 1 KG
SDP

Catalog No. 5000434303
Encompass_Preferred

Tri(2-methylphenyl)phosphine is an organic phosphorus ligand that can be used as a drug intermediate to form complexes with metal salts. It is for research use only and not sold to patients.

  • Molecular weight: 304.37
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, protect from light, stored under nitrogen.
  • In solvent: -80°C for 6 months; -20°C for 1 month (protect from light, stored under nitrogen).
  • Solubility (in vitro): DMSO: 20 mg/mL (65.71 mM; ultrasonic and warming and heat to 60°C; Hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO).

Catalog No. 50-004-34303 Supplier Medchemexpress LLC Supplier No. HY700261KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tri(2-methylphenyl)phosphine is an organic phosphorus ligand that can be used as a drug intermediate to form complexes with metal salts. It is for research use only and not sold to patients.

  • Molecular weight: 304.37
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, protect from light, stored under nitrogen.
  • In solvent: -80°C for 6 months; -20°C for 1 month (protect from light, stored under nitrogen).
  • Solubility (in vitro): DMSO: 20 mg/mL (65.71 mM; ultrasonic and warming and heat to 60°C; Hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.