Learn More
Medchemexpress LLC Tri(2-methylphenyl)phosphine | 6163-58-2 | 99.99% | C21H21P | 100 G
SDP
Catalog No. 5000434302
Encompass_Preferred
Tri(2-methylphenyl)phosphine is an organic phosphorus ligand with potential applications as a drug intermediate. It is primarily used in the formation of complexes with various metal salts and is intended for research purposes only, not for direct patient use.
  • Organic phosphorus ligand
  • Can be used as a drug intermediate
  • Forms complexes with metal salts
  • For research use only
Catalog No. 50-004-34302 Supplier Medchemexpress LLC Supplier No. HY70026100G
Tri(2-methylphenyl)phosphine is an organic phosphorus ligand with potential applications as a drug intermediate. It is primarily used in the formation of complexes with various metal salts and is intended for research purposes only, not for direct patient use.
  • Organic phosphorus ligand
  • Can be used as a drug intermediate
  • Forms complexes with metal salts
  • For research use only