Learn More
Medchemexpress LLC Tri(2-methylphenyl)phosphine | 6163-58-2 | 99.99% | C21H21P | 25 G
SDP
Catalog No. 5000357623
Encompass_Preferred
Tri(2-methylphenyl)phosphine is an organic phosphorus ligand that can be used as a drug intermediate to form complexes with metal salts. It is for research use only.
  • Organic phosphorus ligand
  • Can be used as a drug intermediate
  • Forms complexes with metal salts
  • White to off-white solid appearance
  • For research use only
Catalog No. 50-003-57623 Supplier Medchemexpress LLC Supplier No. 25GHY70026
Tri(2-methylphenyl)phosphine is an organic phosphorus ligand that can be used as a drug intermediate to form complexes with metal salts. It is for research use only.
  • Organic phosphorus ligand
  • Can be used as a drug intermediate
  • Forms complexes with metal salts
  • White to off-white solid appearance
  • For research use only