Learn More
Medchemexpress LLC Tri(Amino-PEG3-amide)-amine | 2523025-40-1 | MFCD28142446 | 98.0% | C33H69N7O12 | 25 MG
SDP
Catalog No. 5000352612
Encompass_Preferred
Tri(Amino-PEG3-amide)-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52612 Supplier Medchemexpress LLC Supplier No. 25MGHY140249
Tri(Amino-PEG3-amide)-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins