Learn More
Medchemexpress LLC Tri(amino-PEG4-amide)-amine | 2523025-41-2 | 95.0% | C39H81N7O15 | 100 MG
SDP
Catalog No. 5000356707
Encompass_Preferred
Tri(Amino-PEG4-amide)-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. These PROTACs consist of two different ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other a specific protein, leveraging the intracellular ubiquitin-proteasome system for selective protein degradation. This product is for research use only.
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • Other ligand targets a specific protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • For research use only
Catalog No. 50-003-56707 Supplier Medchemexpress LLC Supplier No. HY140250100MG
Tri(Amino-PEG4-amide)-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. These PROTACs consist of two different ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other a specific protein, leveraging the intracellular ubiquitin-proteasome system for selective protein degradation. This product is for research use only.
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • Other ligand targets a specific protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • For research use only