Learn More
Medchemexpress LLC Tri(amino-PEG5-amide)-amine | 2055013-52-8 | 99.9% | C45H93N7O18 | 25 MG
SDP
Catalog No. 5000357353
Encompass_Preferred
Tri(amino-PEG5-amide)-amine is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which are molecules designed to selectively degrade target proteins by exploiting the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-57353 Supplier Medchemexpress LLC Supplier No. 25MGHY140251
Tri(amino-PEG5-amide)-amine is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which are molecules designed to selectively degrade target proteins by exploiting the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only