Learn More
Medchemexpress LLC Tri(Amino-PEG5-amide)-amine | 2055013-52-8 | 99.9% | C45H93N7O18 | 50 MG
SDP
Catalog No. 5000357321
Encompass_Preferred
Tri(Amino-PEG5-amide)-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. They consist of two different ligands connected by a linker: one ligand binds to an E3 ubiquitin ligase, and the other to the target protein. This mechanism allows PROTACs to effectively degrade specific proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-57321 Supplier Medchemexpress LLC Supplier No. HY14025150MG
Tri(Amino-PEG5-amide)-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. They consist of two different ligands connected by a linker: one ligand binds to an E3 ubiquitin ligase, and the other to the target protein. This mechanism allows PROTACs to effectively degrade specific proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins