Learn More
Chemglass Life Sciences Tri-Block Reaction Block to fit 1 DRAM, 2 DRAM & 20ML SCINTILLATION VIALS, Black Hard Coated Aluminum
SDP
Catalog No. 501531376
Encompass_Preferred
Tri-Block Reaction Block to fit 1 DRAM, 2 DRAM & 20ML SCINTILLATION VIALS, Black Hard Coated Aluminum. Reaction TRI-BLOCK is designed to fit directly on 135mm diameter circular top hot plate stirrers. Versatile design of the tri-block fits the most common vial sizes on one hot plate: 11ea- 1 Dram vials, 11ea- 2 Dram vials and 7ea 20mL scintillation vials. Tri-block is hard coat anodized black aluminum. A drilled thermowell is provided for insertion of the PT-100 Temperature Sensor.
Catalog No. 50-153-1376 Supplier Chemglass Life Sciences Supplier No. CG1991T10
Tri-Block Reaction Block to fit 1 DRAM, 2 DRAM & 20ML SCINTILLATION VIALS, Black Hard Coated Aluminum. Reaction TRI-BLOCK is designed to fit directly on 135mm diameter circular top hot plate stirrers. Versatile design of the tri-block fits the most common vial sizes on one hot plate: 11ea- 1 Dram vials, 11ea- 2 Dram vials and 7ea 20mL scintillation vials. Tri-block is hard coat anodized black aluminum. A drilled thermowell is provided for insertion of the PT-100 Temperature Sensor.