Learn More
Alpco Ltd Tri-CAT ELISA (Epinephrine, Norepinephrine & Dopamine)
SDP
Supplier:  Alpco Ltd 17TCTHUE03.1
Encompass_Preferred
The Tri-CAT ELISA (Epinephrine, Norepinephrine & Dopamine) is for the quantitative determination of Adrenaline (Epinephrine), Noradrenaline (Norepinephrine), and Dopamine in plasma and urine.
Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3498