Learn More
Description
- Color-keyed buffer preservative (included) prolongs the useful life of the buffer solution
- Dissolve rapidly in water
- Accurate to within ±0.02pH at 25°C
- Each capsule contains sufficient powder to make 100mL of buffer solution
- pH mode: 0.00 to 14.00pH with a resolution of 0.01pH and an accuracy of ±0.01pH
- mV mode: 0 to ±1999 with a resolution of 1mV and an accuracy of ±1mV
- Temperature mode: -5.0° to +105.0°C, resolution: 1°C, accuracy: ±0.5°C
Order Info
Packaged in vials of 10 capsules with internal labels stating pH values.
Specifications
Specifications
| pH | 4, 7, 10 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
