Learn More
Accuflow Systems Steel & O'Brien Tri-Clamp Short Tee, 1/2 in, 316
Supplier:  Accuflow Systems S7MPS.50PL316
Encompass_Preferred
The short outlet tri-clamp tee is a compact BPE fitting for sanitary process lines, providing a short lateral outlet for sampling or routing lines. Fabricated from 316 stainless steel with a polished product-contact finish (PL / SF1), it is intended for bioprocessing and hygienic applications requiring smooth surfaces and quick clamp connections.
  • 316 stainless steel construction for corrosion resistance and process compatibility.
  • Tri-clamp clamp-end connections for quick assembly and sanitary sealing.
  • Polished product-contact finish PL / SF1, 20 μin Ra I.D. and 32 μin Ra O.D.
  • Tube size: 1/2 in (BPE tube O.D.).
  • Dimensions: A 2.375 in (60.33 mm), B 4.75 in (120.65 mm), C 1.000 in (25.4 mm).
  • Compact short-outlet configuration for sampling, instrumentation, or branch routing.
Catalog No. NC3442235