Learn More
Accuflow Systems Steel & O'Brien Concentric Reducer, 316L, 2 x 3/4 In
Catalog No. NC3595632
Encompass_Preferred
Concentric clamp-end reducer for sanitary BPE piping systems. Machined from 316L stainless steel and polished to BPE finishes, this reducer transitions a 2 in tri-clamp connection to a 3/4 in tri-clamp connection and is supplied with material test reports for traceability.
  • 316L stainless steel construction.
  • Sizes: 2 in to 3/4 in concentric configuration.
  • Finish: 20 RA internal (ID), 32 RA external (OD).
  • Clamp (tri-clamp) end connections for sanitary assembly.
  • Meets BPE sanitary specifications.
  • Individually bagged with material test reports (MTRs) and heat lot traceability.
  • Suitable for pharmaceutical and bioprocess piping applications.
Catalog No. NC3595632 Supplier Accuflow Systems Supplier No. S3114MP2X.75PL316
Add to Cart
Add to Cart
Concentric clamp-end reducer for sanitary BPE piping systems. Machined from 316L stainless steel and polished to BPE finishes, this reducer transitions a 2 in tri-clamp connection to a 3/4 in tri-clamp connection and is supplied with material test reports for traceability.
  • 316L stainless steel construction.
  • Sizes: 2 in to 3/4 in concentric configuration.
  • Finish: 20 RA internal (ID), 32 RA external (OD).
  • Clamp (tri-clamp) end connections for sanitary assembly.
  • Meets BPE sanitary specifications.
  • Individually bagged with material test reports (MTRs) and heat lot traceability.
  • Suitable for pharmaceutical and bioprocess piping applications.