Learn More
Accuflow Systems Steel & O'Brien Tri-Clamp Tee, 3/4 in, 316L, Short Outlet
Catalog No. NC3442238
Encompass_Preferred
The Steel & O'Brien Tri-Clamp Short Outlet Tee is a BPE sanitary fitting designed for hygienic process lines. The short outlet branch reduces dead space at instrumentation and sampling points, while clamp-end connections enable rapid assembly and disassembly for cleaning and maintenance. The 316L stainless steel construction and PL (SF1) surface finish provide smooth internal surfaces suitable for pharmaceutical, biotech, and food-processing applications.
  • Short outlet tee configuration reduces dead space in process lines.
  • Clamp end connections for tri-clamp quick assembly and disassembly.
  • 316L stainless steel construction for corrosion resistance and cleanability.
  • PL (SF1) surface finish with typical Ra values: ID ~20 μin, OD ~32 μin.
  • Designed for BPE sanitary piping and hygienic process applications.
  • Commonly used for sampling, instrumentation, and sanitary branch connections.
Catalog No. NC3442238 Supplier Accuflow Systems Supplier No. S7MPS.75PL316
The Steel & O'Brien Tri-Clamp Short Outlet Tee is a BPE sanitary fitting designed for hygienic process lines. The short outlet branch reduces dead space at instrumentation and sampling points, while clamp-end connections enable rapid assembly and disassembly for cleaning and maintenance. The 316L stainless steel construction and PL (SF1) surface finish provide smooth internal surfaces suitable for pharmaceutical, biotech, and food-processing applications.
  • Short outlet tee configuration reduces dead space in process lines.
  • Clamp end connections for tri-clamp quick assembly and disassembly.
  • 316L stainless steel construction for corrosion resistance and cleanability.
  • PL (SF1) surface finish with typical Ra values: ID ~20 μin, OD ~32 μin.
  • Designed for BPE sanitary piping and hygienic process applications.
  • Commonly used for sampling, instrumentation, and sanitary branch connections.