Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TECHLAB™ TRI-COMBO PARASITE SCREEN

Catalog No. 23900557
Change view
Click to view available options
Quantity:
96/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-900-557 96/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-900-557 Supplier TECHLAB™ Supplier No. T30408
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The TRI-COMBO PARASITE SCREEN test is an enzyme immunoassay for the simultaneous qualitative detection of Giardia spp., Cryptosporidium spp., and/or E. histolytica antigen in human fecal specimens.

The test is indicated for use as a screen for fecal specimens from patients with gastrointestinal illness including dysentery to aid in the diagnosis of enteric infection resulting from giardiasis, cryptosporidiosis or amebiasis.

The TRI-COMBO PARASITE SCREEN test serves as a screen to rule out negative specimens and reduce the number of specimens that require additional follow-up testing. The test uses monoclonal and polyclonal antibodies to cell-surface antigens of Giardia, Cryptosporidium, and E. histolytica.

  • Ideal screening test for the three most common parasitic causes of diarrhea

Specifications

Type Monoclonal ELISA
For Use With (Application) Detecting Giardia, Cryptosporidium and E. histolytica Antigens in Fecal Specimens
No. of Reactions 96 Tests
Quantity 96/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.