Learn More
TWD Scientific LLC Beaker, Tri-Cornered, Twdbiox, 250Ml Polypropylene, Disposable/Reusable, 3 Pour Spout, Molded Graduations, Autoclavable, 100/Case.
SDP
Catalog No. 502285102 Shop All TWD Scientific LLC Products
Encompass_Preferred
250Ml Clear Polypropylene (Disposable/Reusable) Tri-Cornered Beakers With Three Pour Spouts And Molded Graduations. Beakers Are Autoclavable. 25 Beakers/Pack, 4 Packs/Case, Price/Case
This item is not returnable. View return policy
Catalog No. 50-228-5102 Supplier TWD Scientific LLC Supplier No. BX930250
Add to Cart
Add to Cart
250Ml Clear Polypropylene (Disposable/Reusable) Tri-Cornered Beakers With Three Pour Spouts And Molded Graduations. Beakers Are Autoclavable. 25 Beakers/Pack, 4 Packs/Case, Price/Case