Learn More
TWD Scientific LLC Beaker, Tri-Cornered, Twdbiox, 400Ml Polypropylene, Disposable/Reusable, 3 Pour Spout, Molded Graduations, Autoclavable, 100/Case.
SDP
Catalog No. 502285103 Shop All TWD Scientific LLC Products
Encompass_Preferred
400Ml Clear Polypropylene (Disposable/Reusable) Tri-Cornered Beakers With Three Pour Spouts And Molded Graduations. Beakers Are Autoclavable. 25 Beakers/Pack, 4 Packs/Case, Price/Case
This item is not returnable. View return policy
Catalog No. 50-228-5103 Supplier TWD Scientific LLC Supplier No. BX930400
400Ml Clear Polypropylene (Disposable/Reusable) Tri-Cornered Beakers With Three Pour Spouts And Molded Graduations. Beakers Are Autoclavable. 25 Beakers/Pack, 4 Packs/Case, Price/Case