Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tri(ethylene glycol) divinyl ether

Catalog No. 3298001L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-980-01L 1 L
32-980-0250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 32-980-01L Supplier Sigma Aldrich Supplier No. 3298001L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tri(ethylene glycol) divinyl ether is a bifunctional monomer that can be used in a variety of properties such as: formation of gel polymer electrolyte for lithium ion batteries, for the investigation of photo-initiators by cationic polymerization of vinyl ester monomers

Specifications

CAS 765-12-8
Density 0.99 g/mL (at 25°C)
Boiling Point 120°C to 126°C (18 mmHg)
Molecular Formula C10H18O4
Refractive Index n20/D 1.453 (literature)
Linear Formula H2C=CH(OCH2CH2)3 OCH=CH2
Quantity 1 L
MDL Number MFCD00074891
Molecular Weight (g/mol) 202.25
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.