Learn More
Grainger TRI-FLOW General Purpose Lubricant, Mineral Oil, PTFE, 12 oz
Catalog No. NC0976042
Encompass_Preferred
This high-performance lubricant features a concentrated mineral oil formula containing micro-sized particles to deliver advanced protection. It is designed to penetrate inaccessible areas while effectively cleaning and displacing contaminants. By depositing a long-lasting wet film, it provides a protective shield that inhibits the formation of rust and corrosive buildup, helping to reduce friction and wear while extending the service life of critical mechanisms.
  • Effective operating temperature range of -60°F to 475°F
  • Contains PTFE for enhanced lubrication performance
  • Provides a protective shield that resists rust and corrosion
  • Rejects dirt, dust, sand, and abrasive particles
  • Increases part life and reduces costly equipment breakdowns
  • NSF registered #127540 (Category Code H2)
  • 50 state VOC compliant
Catalog No. NC0976042 Supplier Grainger Supplier No. 3KB67
This high-performance lubricant features a concentrated mineral oil formula containing micro-sized particles to deliver advanced protection. It is designed to penetrate inaccessible areas while effectively cleaning and displacing contaminants. By depositing a long-lasting wet film, it provides a protective shield that inhibits the formation of rust and corrosive buildup, helping to reduce friction and wear while extending the service life of critical mechanisms.
  • Effective operating temperature range of -60°F to 475°F
  • Contains PTFE for enhanced lubrication performance
  • Provides a protective shield that resists rust and corrosion
  • Rejects dirt, dust, sand, and abrasive particles
  • Increases part life and reduces costly equipment breakdowns
  • NSF registered #127540 (Category Code H2)
  • 50 state VOC compliant