Learn More
Medchemexpress LLC Tri-GalNAc biotin | 00-00-0 | 99.8% | C83H146N14O34S | 10MG
SDP
Catalog No. 5000261602
Encompass_Preferred
tri-GalNAc biotin is a biotinylated asialoglycoprotein receptor (ASGPR) ligand used for lysosome-targeting chimera (LYTAC) research. It promotes ASGPR-mediated cellular uptake of neutravidin and delivers bound protein to lysosomes for degradation. Supplied as a white to off-white solid, it is highly pure and suitable for in vitro and in vivo formulation studies.
  • Biotinylated ASGPR ligand for lysosomal targeting studies.
  • Promotes receptor-mediated uptake of neutravidin into cells.
  • High purity suitable for research applications.
  • Soluble in DMSO (100 mg/mL) for in vitro use.
  • Formulatable in recommended in vivo vehicles at ≥2.5 mg/mL.
  • Stable as a powder at -20°C for long-term storage.
Catalog No. 50-002-61602 Supplier Medchemexpress LLC Supplier No. HY14710210MG
tri-GalNAc biotin is a biotinylated asialoglycoprotein receptor (ASGPR) ligand used for lysosome-targeting chimera (LYTAC) research. It promotes ASGPR-mediated cellular uptake of neutravidin and delivers bound protein to lysosomes for degradation. Supplied as a white to off-white solid, it is highly pure and suitable for in vitro and in vivo formulation studies.
  • Biotinylated ASGPR ligand for lysosomal targeting studies.
  • Promotes receptor-mediated uptake of neutravidin into cells.
  • High purity suitable for research applications.
  • Soluble in DMSO (100 mg/mL) for in vitro use.
  • Formulatable in recommended in vivo vehicles at ≥2.5 mg/mL.
  • Stable as a powder at -20°C for long-term storage.