Learn More
Medchemexpress LLC Tri-GalNAc biotin | 3062597-46-7 | 99.8% | 1916.19 | 5 MG
SDP
Catalog No. 5000359923
Encompass_Preferred
Tri-GalNAc biotin is a biotinylated ASGPR (Asialoglycoprotein receptor) ligand. It promotes cellular uptake of neutravidin (NA) via ASGPR and delivers neutravidin to lysosomes for degradation. This compound can be used for research of LYsosome TArgeting Chimera (LYTAC).
  • Promotes cellular uptake of neutravidin (NA) via ASGPR.
  • Delivers neutravidin to lysosomes for degradation.
  • Facilitates ASGPR-dependent cellular uptake of fluorescently labeled NeutrAvidin (NA-650).
  • Useful for research of LYsosome TArgeting Chimera (LYTAC).
Catalog No. 50-003-59923 Supplier Medchemexpress LLC Supplier No. HY1471025MG
Tri-GalNAc biotin is a biotinylated ASGPR (Asialoglycoprotein receptor) ligand. It promotes cellular uptake of neutravidin (NA) via ASGPR and delivers neutravidin to lysosomes for degradation. This compound can be used for research of LYsosome TArgeting Chimera (LYTAC).
  • Promotes cellular uptake of neutravidin (NA) via ASGPR.
  • Delivers neutravidin to lysosomes for degradation.
  • Facilitates ASGPR-dependent cellular uptake of fluorescently labeled NeutrAvidin (NA-650).
  • Useful for research of LYsosome TArgeting Chimera (LYTAC).